Please login to your online account to display your discounted pricing

anti-Rat AA-NAT (residues 198-208) Synthetic, Thermo Scientific™

Rt AA-NAT (residues 198-208) Synthetic peptide PEP-065 from Thermo Scientific™ detects Rt AA-NAT (residues 198-208) in Not Applicable samples, and is validated for Control

$158.05 - $158.05

Specifications

Concentration 0.5mg/mL
Quantity 50μg
Format Purified
For Use With (Application) Control
Lyophilized Yes
View More Specs
Products
Catalog Number Mfr. No. Product Type Sequence Price Quantity    

PEP063

 
pierce biotechnology inc
PEP063
Rat AA-Nat (Residues 24-33) Synthetic Peptide C24-Q-R-R-H-T-L-P-A-S33 Each for $158.05

PEP065

 
pierce biotechnology inc
PEP065
Rat AA-NAT (Residues 198-208) Synthetic Peptide C198-H-T-F-L-R-R-N-S-G-C208 Each for $158.05
Description & Specifications

Specifications

Concentration 0.5mg/mL
Quantity 50μg
Format Purified
For Use With (Application) Control
Lyophilized Yes
Purity >95%
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Rt AA-NAT (residues 198-208) Synthetic peptide PEP-065 from Thermo Scientific™ detects Rt AA-NAT (residues 198-208) in Not Applicable samples, and is validated for Control

PEP-065 is a 11 amino acid synthetic peptide whose sequence corresponds to residues 198-208 of rat AA-NAT. The sequence of this peptide is (amino to carboxy terminus): C198 - H - T - F - L - R - R - N - S - G - C208
Host Species: N/A
Cross Reactivity: Not Applicable
Formulation: 50μg purified Peptide (0.5mg/mL) no preservative;

PEP-065 detects Rat AA-NAT (residues 198-208) in Not Applicable samples. PEP-065 has been successfully used in Control procedures. PEP-065