Please login to your online account to display your discounted pricing

anti-Human BACE2 (residues 441-457) Synthetic, Thermo Scientific™


Hu BACE2 (residues 441-457) Synthetic peptide PEP-166 from Thermo Scientific™ detects Hu BACE2 (residues 441-457) in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  PEP166

 View more versions of this product

Catalog No. PEP166

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control
Format Purified
Formulation 50μg purified peptide (0.5mg/mL) and no preservative
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Human BACE2 (Residues 441-457) Synthetic Peptide
Concentration 0.5mg/mL
Lyophilized Yes
Sequence V(441)-S-E-I-S-G-P-F-S-T-E-D-I-A-S-N-C(457)
Quantity 50μg
Purity >95%

Hu BACE2 (residues 441-457) Synthetic peptide PEP-166 from Thermo Scientific™ detects Hu BACE2 (residues 441-457) in Not Applicable samples, and is validated for Control


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-166 is an 18 amino acid synthetic peptide whose sequence is derived from human BACE2 protein. The sequence of this peptide is (amino to carboxy terminus): V (441)-S-E-I-S-G-P-F-S-T-E-D-I-A-S-N-C (457) This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human BACE2, catalog # PA1-753. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.