Please login to your online account to display your discounted pricing

anti-Human BACE2 (residues 441-457) Synthetic, Thermo Scientific™

Hu BACE2 (residues 441-457) Synthetic peptide PEP-166 from Thermo Scientific™ detects Hu BACE2 (residues 441-457) in Not Applicable samples, and is validated for Control

$144.64 - $158.05

Specifications

Concentration 0.5mg/mL
Quantity 50μg
Format Purified
For Use With (Application) Control
Lyophilized Yes
View More Specs
Products
Catalog Number Mfr. No. Product Type Sequence Price Quantity    

PEP167

 
pierce biotechnology inc
PEP167
Human BACE2 (Residues 503-518) Synthetic Peptide C-P(503)-E-V-V-N-D-E-S-S-L-V-R-H-R-W-K(518) Each for $158.05

PEP166

 
pierce biotechnology inc
PEP166
Human BACE2 (Residues 441-457) Synthetic Peptide V(441)-S-E-I-S-G-P-F-S-T-E-D-I-A-S-N-C(457) Each for $144.64
Description & Specifications

Specifications

Concentration 0.5mg/mL
Quantity 50μg
Format Purified
For Use With (Application) Control
Lyophilized Yes
Purity >95%
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Hu BACE2 (residues 441-457) Synthetic peptide PEP-166 from Thermo Scientific™ detects Hu BACE2 (residues 441-457) in Not Applicable samples, and is validated for Control

PEP-166 is an 18 amino acid synthetic peptide whose sequence is derived from human BACE2 protein. The sequence of this peptide is (amino to carboxy terminus): V(441) - S - E - I - S - G - P - F - S - T - E - D - I - A - S - N - C(457)
Host Species: N/A
Cross Reactivity: Not Applicable
Formulation: 50μg purified Peptide (0.5mg/mL) no preservative;

PEP-166 detects Human BACE2 (residues 441-457) in Not Applicable samples. PEP-166 has been successfully used in Control procedures. PEP-166