Please login to your online account to display your discounted pricing

anti-Rat and Human SAP97 (residues 16-33) Synthetic, Thermo Scientific™


Rt and Hu SAP97 (residues 16-33) Synthetic peptide PEP-022 detects Rt and Hu SAP97 (residues 16-33) in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  PEP022

 View more versions of this product

Catalog No. PEP022

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control
Format Purified
Formulation 50μg purified peptide (0.5mg/mL) and no preservative
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Rat and Human SAP97 (Residues 16-33) Synthetic Peptide
Concentration 0.5mg/mL
Lyophilized Yes
Sequence E16-E-Y-R-S-K-L-S-Q-T-E-D-R-Q-L-R-S-S33
Purity >95%
Quantity 50μg

Rt and Hu SAP97 (residues 16-33) Synthetic peptide PEP-022 detects Rt and Hu SAP97 (residues 16-33) in Not Applicable samples, and is validated for Control


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-022 is an 18 amino acid (amino acids 16-33) synthetic peptide whose sequence is found in rat and human SAP97. The sequence of this peptide is (amino to carboxy terminus): E16-E-Y-R-S-K-L-S-Q-T-E-D-R-Q-L-R-S-S33 This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and SAP97, catalog # PA1-044. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody. Reconstitute with 0.1 ml of distilled water.