Please login to your online account to display your discounted pricing

anti-Rat and Human SAP97 (residues 16-33) Synthetic, Thermo Scientific™

Rt and Hu SAP97 (residues 16-33) Synthetic peptide PEP-022 detects Rt and Hu SAP97 (residues 16-33) in Not Applicable samples, and is validated for Control

$158.05 - $158.05

Specifications

Concentration 0.5mg/mL
Quantity 50μg
Format Purified
For Use With (Application) Control
Lyophilized Yes
View More Specs
Products
Catalog Number Mfr. No. Product Type Sequence Price Quantity    

PEP022

 
pierce biotechnology inc
PEP022
Rat and Human SAP97 (Residues 16-33) Synthetic Peptide E16-E-Y-R-S-K-L-S-Q-T-E-D-R-Q-L-R-S-S33 Each for $158.05

PEP055

 
pierce biotechnology inc
PEP055
Rat SAP97 (Residues 115-133) Synthetic Peptide V115-L-P-S-E-R-I-S-P-Q-V-P-N-E-V-L-G-P-E133 (C) Each for $158.05
Description & Specifications

Specifications

Concentration 0.5mg/mL
Quantity 50μg
Format Purified
For Use With (Application) Control
Lyophilized Yes
Purity >95%
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Rt and Hu SAP97 (residues 16-33) Synthetic peptide PEP-022 detects Rt and Hu SAP97 (residues 16-33) in Not Applicable samples, and is validated for Control

PEP-022 is an 18 amino acid (amino acids 16-33) synthetic peptide whose sequence is found in rat and human SAP97. The sequence of this peptide is (amino to carboxy terminus): E16 - E - Y - R - S - K - L - S - Q - T - E - D - R - Q - L - R - S - S33
Host Species: N/A
Cross Reactivity: Not Applicable
Formulation: 50μg purified Peptide (0.5mg/mL) no preservative;

PEP-022 detects Rat and Human SAP97 (residues 16-33) in Not Applicable samples. PEP-022 has been successfully used in Control procedures. PEP-022