Please login to your online account to display your discounted pricing

anti-Rat and Human SAP97 (residues 16-33) Synthetic, Thermo Scientific™


Rt and Hu SAP97 (residues 16-33) Synthetic peptide PEP-022 detects Rt and Hu SAP97 (residues 16-33) in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  PEP055

 View more versions of this product

Catalog No. PEP055

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control
Format Purified
Formulation 50μg purified peptide (0.5mg/mL) and no preservative
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Rat SAP97 (Residues 115-133) Synthetic Peptide
Quantity 50μg
Concentration 0.5mg/mL
Lyophilized Yes
Sequence V115-L-P-S-E-R-I-S-P-Q-V-P-N-E-V-L-G-P-E133 (C)
Purity >95%

Rt and Hu SAP97 (residues 16-33) Synthetic peptide PEP-022 detects Rt and Hu SAP97 (residues 16-33) in Not Applicable samples, and is validated for Control


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-055 is a 20 amino acid synthetic peptide whose sequence corresponds to residues 115-133 of rat SAP97. The sequence of this peptide is (amino to carboxy terminus): V115-L-P-S-E-R-I-S-P-Q-V-P-N-E-V-L-G-P-E133 (C) (The C-terminal cysteine has been added to facilitate conjugation and is not a part of the native rat SAP97 sequence). This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and SAP97, catalog # PA1-741. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.