Please login to your online account to display your discounted pricing

anti-Mouse Resistin (residues 21-39) Synthetic, Thermo Scientific™


Ms Resistin (residues 21-39) Synthetic peptide PEP-165 from Thermo Scientific™ detects Ms Resistin (residues 21-39) in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  PEP165

 View more versions of this product

Catalog No. PEP165

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control
Format Purified
Formulation 50μg purified peptide (0.5mg/mL) and no preservative
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Mouse Resistin (Residues 21-39) Synthetic Peptide
Lyophilized Yes
Purity >70%
Quantity 50μg
Concentration 0.5mg/mL
Sequence S(21)-S-M-P-L-C-P-I-D-E-A-I-D-K-K-I-K-Q-D-C(39)

Ms Resistin (residues 21-39) Synthetic peptide PEP-165 from Thermo Scientific™ detects Ms Resistin (residues 21-39) in Not Applicable samples, and is validated for Control


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-165 is an 18 amino acid synthetic peptide whose sequences are derived from mouse resistin protein. The sequence of this peptide is (amino to carboxy terminus): S (21)-S-M-P-L-C-P-I-D-E-A-I-D-K-K-I-K-Q-D-C (39) This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and mouse resistin, catalog # PA1-1050. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.