Please login to your online account to display your discounted pricing

anti-Mouse Resistin (residues 21-39) Synthetic, Thermo Scientific™

Ms Resistin (residues 21-39) Synthetic peptide PEP-165 from Thermo Scientific™ detects Ms Resistin (residues 21-39) in Not Applicable samples, and is validated for Control

$158.05 - $158.05

Specifications

Concentration 0.5mg/mL
Quantity 50μg
Format Purified
For Use With (Application) Control
Lyophilized Yes
View More Specs
Products
Catalog Number Mfr. No. Product Type Sequence Price Quantity    

PEP165

 
pierce biotechnology inc
PEP165
Mouse Resistin (Residues 21-39) Synthetic Peptide S(21)-S-M-P-L-C-P-I-D-E-A-I-D-K-K-I-K-Q-D-C(39) Each for $158.05

PEP218

 
pierce biotechnology inc
PEP218
Resistin (Residues 22-39) Synthetic Peptide C(22) S M E E A I N E R I Q E V A G S L(39) Each for $158.05
Description & Specifications

Specifications

Concentration 0.5mg/mL
Quantity 50μg
Format Purified
For Use With (Application) Control
Lyophilized Yes
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Ms Resistin (residues 21-39) Synthetic peptide PEP-165 from Thermo Scientific™ detects Ms Resistin (residues 21-39) in Not Applicable samples, and is validated for Control

PEP-165 is an 18 amino acid synthetic peptide whose sequences are derived from mouse resistin protein. The sequence of this peptide is (amino to carboxy terminus): S(21) - S - M - P - L - C - P - I - D - E - A - I - D - K - K - I - K - Q - D - C(39)
Host Species: N/A
Cross Reactivity: Not Applicable
Formulation: 50μg purified Peptide (0.5mg/mL) no preservative;

PEP-165 detects Mouse Resistin (residues 21-39) in Not Applicable samples. PEP-165 has been successfully used in Control procedures. PEP-165