Please login to your online account to display your discounted pricing

anti-Mouse Resistin (residues 21-39) Synthetic, Thermo Scientific™


Ms Resistin (residues 21-39) Synthetic peptide PEP-165 from Thermo Scientific™ detects Ms Resistin (residues 21-39) in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  PEP218

 View more versions of this product

Catalog No. PEP218

  • / Each
 

Description & Specifications

Specifications

Formulation 50μg purified peptide (0.5mg/mL) and no preservative
Format Purified
For Use With (Application) Control
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Resistin (Residues 22-39) Synthetic Peptide
Quantity 50μg
Concentration 0.5mg/mL
Sequence C(22) S M E E A I N E R I Q E V A G S L(39)
Lyophilized Yes
Purity >95%

Ms Resistin (residues 21-39) Synthetic peptide PEP-165 from Thermo Scientific™ detects Ms Resistin (residues 21-39) in Not Applicable samples, and is validated for Control


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-218 is a 18 amino acid synthetic peptide. The sequence of this peptide is (amino to carboxy terminus: C (22) S M E E A I N E R I Q E V A G S L (39). This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA1-1049. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.