Please login to your online account to display your discounted pricing

anti-Mouse MeCP2 (residues 1-15) Synthetic, Thermo Scientific™


Ms MeCP2 (residues 1-15) Synthetic peptide PEP-120 from Thermo Scientific™ specifically detects Ms MeCP2 (residues 1-15) in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  PEP121

 View more versions of this product

Catalog No. PEP121

  • / Each
 

Description & Specifications

Specifications

Formulation 50μg purified peptide (0.5mg/mL) and no preservative
Format Purified
For Use With (Application) Control
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Mouse MeCP2 (Residues 469-486) Synthetic Peptide
Sequence C-P(469)-N-R-E-E-P-V-D-S-R-T-P-V-T-E-R-V-S(486)
Concentration 0.5mg/mL
Lyophilized Yes
Purity >95%
Quantity 50μg

Ms MeCP2 (residues 1-15) Synthetic peptide PEP-120 from Thermo Scientific™ specifically detects Ms MeCP2 (residues 1-15) in Not Applicable samples, and is validated for Control


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-121 is a 18 amino acid synthetic peptide whose sequences are derived from amino acids 469-486 of mouse MeCP2 protein with a amino-terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): C-P (469)-N-R-E-E-P-V-D-S-R-T-P-V-T-E-R-V-S (486) PEP-121 may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and mouse MeCP2, catalog # PA1-888. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.