Please login to your online account to display your discounted pricing

anti-Mouse MeCP2 (residues 1-15) Synthetic, Thermo Scientific™


Ms MeCP2 (residues 1-15) Synthetic peptide PEP-120 from Thermo Scientific™ specifically detects Ms MeCP2 (residues 1-15) in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  PEP120

 View more versions of this product

Catalog No. PEP120

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control
Format Purified
Formulation 50μg purified peptide (0.5mg/mL) and no preservative
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Mouse MeCP2 (Residues 1-15) Synthetic Peptide
Concentration 0.5mg/mL
Lyophilized Yes
Sequence M(1)-V-A-G-M-L-G-L-R-E-E-K-S-E-D(15)-C
Quantity 50μg
Purity >95%

Ms MeCP2 (residues 1-15) Synthetic peptide PEP-120 from Thermo Scientific™ specifically detects Ms MeCP2 (residues 1-15) in Not Applicable samples, and is validated for Control


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-120 is a 15 amino acid synthetic peptide whose sequences are derived from amino acids 1-15 of mouse MeCP2 protein with a carboxy-terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): M (1)-V-A-G-M-L-G-L-R-E-E-K-S-E-D (15)-C This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and mouse MeCP2, catalog # PA1-887. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.