Please login to your online account to display your discounted pricing

anti-Mouse MeCP2 (residues 1-15) Synthetic, Thermo Scientific™

Ms MeCP2 (residues 1-15) Synthetic peptide PEP-120 from Thermo Scientific™ specifically detects Ms MeCP2 (residues 1-15) in Not Applicable samples, and is validated for Control

$142.10 - $144.64

Specifications

Concentration 0.5mg/mL
Quantity 50μg
Format Purified
For Use With (Application) Control
Lyophilized Yes
View More Specs
Products
Catalog Number Mfr. No. Product Type Sequence Price Quantity    

PEP120

 
pierce biotechnology inc
PEP120
Mouse MeCP2 (Residues 1-15) Synthetic Peptide M(1)-V-A-G-M-L-G-L-R-E-E-K-S-E-D(15)-C Each for $144.64

PEP121

 
pierce biotechnology inc
PEP121
Mouse MeCP2 (Residues 469-486) Synthetic Peptide C-P(469)-N-R-E-E-P-V-D-S-R-T-P-V-T-E-R-V-S(486) Each for $142.10
Description & Specifications

Specifications

Concentration 0.5mg/mL
Quantity 50μg
Format Purified
For Use With (Application) Control
Lyophilized Yes
Purity >95%
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Ms MeCP2 (residues 1-15) Synthetic peptide PEP-120 from Thermo Scientific™ specifically detects Ms MeCP2 (residues 1-15) in Not Applicable samples, and is validated for Control

PEP-120 is a 15 amino acid synthetic peptide whose sequences are derived from amino acids 1-15 of mouse MeCP2 protein with a carboxy-terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): M(1) - V - A - G - M - L - G - L - R - E - E - K - S - E - D(15) - C
Host Species: N/A
Cross Reactivity: Not Applicable
Formulation: 50μg purified Peptide (0.5mg/mL) no preservative;

PEP-120 detects Mouse MeCP2 (residues 1-15) in Not Applicable samples. PEP-120 has been successfully used in Control procedures. PEP-120