Please login to your online account to display your discounted pricing

anti-Human IRS-1 (residues 301-315) Synthetic, Thermo Scientific™


Hu IRS-1 (residues 301-315) Synthetic peptide PEP-191 from Thermo Scientific™ detects Hu IRS-1 (residues 301-315) in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  PEP192

 View more versions of this product

Catalog No. PEP192

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control
Format Purified
Formulation 35μg purified peptide (0.35mg/mL) and no preservative
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Human IRS-1 (Residues 602-612) Synthetic Peptide
Quantity 35μg
Concentration 0.35mg/mL
Lyophilized Yes
Sequence L H T D D G Y M P M S P G V(NH2)
Purity >95%

Hu IRS-1 (residues 301-315) Synthetic peptide PEP-191 from Thermo Scientific™ detects Hu IRS-1 (residues 301-315) in Not Applicable samples, and is validated for Control


Formulation: 35μg purified peptide (0.35mg/mL) and no preservative
PEP-192 is a 14 amino acid synthetic peptide corresponding to amino acid residues 602-615 from human IRS-1. The sequence of this peptide is (amino to carboxy terminus): L H T D D G Y M P M S P G V (NH2). This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA1-1058. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.