Please login to your online account to display your discounted pricing

anti-Human IRS-1 (residues 301-315) Synthetic, Thermo Scientific™


Hu IRS-1 (residues 301-315) Synthetic peptide PEP-191 from Thermo Scientific™ detects Hu IRS-1 (residues 301-315) in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  PEP191

 View more versions of this product

Catalog No. PEP191

  • / Each
 

Description & Specifications

Specifications

Formulation 50μg purified peptide (0.5mg/mL) and no preservative
For Use With (Application) Control
Format Purified
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Human IRS-1 (Residues 301-315) Synthetic Peptide
Sequence E S I T A T S P A S M V G G K(NH2)
Purity >95%
Concentration 0.5mg/mL
Lyophilized Yes
Quantity 50μg

Hu IRS-1 (residues 301-315) Synthetic peptide PEP-191 from Thermo Scientific™ detects Hu IRS-1 (residues 301-315) in Not Applicable samples, and is validated for Control


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-191 is a 15 amino acid synthetic peptide corresponding to amino acid residues 301-315 from human IRS-1. The sequence of this peptide is (amino to carboxy terminus): E S I T A T S P A S M V G G K (NH2). This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA1-1057. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.