Please login to your online account to display your discounted pricing

anti-Human IRS-1 (residues 301-315) Synthetic, Thermo Scientific™

Hu IRS-1 (residues 301-315) Synthetic peptide PEP-191 from Thermo Scientific™ detects Hu IRS-1 (residues 301-315) in Not Applicable samples, and is validated for Control

$144.64 - $158.05

Specifications

Format Purified
For Use With (Application) Control
Lyophilized Yes
Purity >95%
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
View More Specs
Products
Catalog Number Mfr. No. Concentration Product Type Sequence Quantity Price Quantity    

PEP191

 
pierce biotechnology inc
PEP191
0.5mg/mL Human IRS-1 (Residues 301-315) Synthetic Peptide E S I T A T S P A S M V G G K(NH2) 50μg Each for $144.64

PEP192

 
pierce biotechnology inc
PEP192
0.35mg/mL Human IRS-1 (Residues 602-612) Synthetic Peptide L H T D D G Y M P M S P G V(NH2) 35μg Each for $158.05

PEP193

 
pierce biotechnology inc
PEP193
0.5mg/mL Human IRS-1 (Residues 631-646) Synthetic Peptide D Y M P M S P K S V S A P Q Q I(NH2) 50μg Each for $144.64
Description & Specifications

Specifications

Format Purified
For Use With (Application) Control
Lyophilized Yes
Purity >95%
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Hu IRS-1 (residues 301-315) Synthetic peptide PEP-191 from Thermo Scientific™ detects Hu IRS-1 (residues 301-315) in Not Applicable samples, and is validated for Control

PEP-191 is a 15 amino acid synthetic peptide corresponding to amino acid residues 301-315 from human IRS-1. The sequence of this peptide is (amino to carboxy terminus): E S I T A T S P A S M V G G K(NH2).
Host Species: N/A
Cross Reactivity: Not Applicable
Formulation: 50μg purified Peptide (0.5mg/mL) no preservative;

PEP-191 detects Human IRS-1 (residues 301-315) in Not Applicable samples. PEP-191 has been successfully used in Control procedures. PEP-191