Please login to your online account to display your discounted pricing

anti-Mouse CLOCK (residues 11-30) Synthetic, Thermo Scientific™


Ms CLOCK (residues 11-30) Synthetic peptide PEP-072 from Thermo Scientific™ specifically detects Ms CLOCK (residues 11-30) in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  PEP073

 View more versions of this product

Catalog No. PEP073

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control
Formulation 50μg purified peptide (0.5mg/mL) and no preservative
Format Purified
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Mouse CLOCK (Residues 394-410) Synthetic Peptide
Quantity 50μg
Sequence (C)-S394-L-P-E-T-A-A-D-K-S-Q-D-S-G-S-D-N-410
Concentration 0.5mg/mL
Lyophilized Yes
Purity >95%

Ms CLOCK (residues 11-30) Synthetic peptide PEP-072 from Thermo Scientific™ specifically detects Ms CLOCK (residues 11-30) in Not Applicable samples, and is validated for Control


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-073 is a 17 amino acid synthetic peptide whose sequence corresponds to residues 394-410 of mouse CLOCK with an amino terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): C S (394) L P E T A A D K S Q D S G S D N (410) This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and mouse CLOCK, catalog # PA1-521. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.