missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse CLOCK (residues 11-30) Synthetic Peptide, Invitrogen™

Mouse CLOCK (residues 11-30) Synthetic Peptide, PEP-072, from Invitrogen™

Manufacturer:  Pierce Biotechnology Inc PEP072

Catalog No. PEP072


 

Specifications

Description

Mouse CLOCK (residues 11-30) Synthetic Peptide, PEP-072, from Invitrogen.


Formulation: 50μg purified peptide and no preservative
PEP-072 is a 20 amino acid synthetic peptide whose sequence corresponds to residues 11-30 of mouse CLOCK with an amino terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): S11-S-I-V-D-R-D-D-S-S-I-F-D-G-L-V-E-E-D-D30-C This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and mouse CLOCK, catalog # PA1-520. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.
Description & Specifications

Specifications

>95%
0.35mg/mL
Purified
Yes
store at -20°C, Avoid Freeze/Thaw Cycles
50μg
Control
50μg purified peptide and no preservative
S11-S-I-V-D-R-D-D-S-S-I-F-D-G-L-V-E-E-D-D30-C
Mouse CLOCK (Residues 11-30) Synthetic Peptide