Please login to your online account to display your discounted pricing

anti-Mouse CLOCK (residues 11-30) Synthetic, Thermo Scientific™

Ms CLOCK (residues 11-30) Synthetic peptide PEP-072 from Thermo Scientific™ specifically detects Ms CLOCK (residues 11-30) in Not Applicable samples, and is validated for Control

$142.10 - $158.05

Specifications

Format Purified
For Use With (Application) Control
Lyophilized Yes
Purity >95%
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
View More Specs
Products
Catalog Number Mfr. No. Concentration Product Type Sequence Quantity Price Quantity    

PEP072

 
pierce biotechnology inc
PEP072
0.35mg/mL Mouse CLOCK (Residues 11-30) Synthetic Peptide S11-S-I-V-D-R-D-D-S-S-I-F-D-G-L-V-E-E-D-D30-C 50μg Each for $142.10

PEP073

 
pierce biotechnology inc
PEP073
0.5mg/mL Mouse CLOCK (Residues 394-410) Synthetic Peptide (C)-S394-L-P-E-T-A-A-D-K-S-Q-D-S-G-S-D-N-410 50μg Each for $158.05
Description & Specifications

Specifications

Format Purified
For Use With (Application) Control
Lyophilized Yes
Purity >95%
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Ms CLOCK (residues 11-30) Synthetic peptide PEP-072 from Thermo Scientific™ specifically detects Ms CLOCK (residues 11-30) in Not Applicable samples, and is validated for Control

PEP-072 is a 20 amino acid synthetic peptide whose sequence corresponds to residues 11-30 of mouse CLOCK with an amino terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): S11 - S - I - V - D - R - D - D - S - S - I - F - D - G - L - V - E - E - D - D30 - C
Host Species: N/A
Cross Reactivity: Not Applicable
Formulation: 50μg purified Peptide no preservative;

PEP-072 detects Mouse CLOCK (residues 11-30) in Not Applicable samples. PEP-072 has been successfully used in Control procedures. PEP-072