Please login to your online account to display your discounted pricing

anti-Mouse Calreticulin (residues 24-43) Synthetic, Thermo Scientific™


Ms Calreticulin (residues 24-43) Synthetic peptide PEP-018 detects Ms Calreticulin (residues 24-43) in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  PEP018

 View more versions of this product

Catalog No. PEP018

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control
Format Purified
Formulation 50μg purified peptide (0.5mg/mL) and no preservative
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Mouse Calreticulin (Residues 24-43) Synthetic Peptide
Lyophilized Yes
Sequence K-E -Q-F-L-D-G-D-A-W-T-N-R-W-V-E-S-K-H-K-C
Purity >95%
Concentration 0.5mg/mL
Quantity 50μg

Ms Calreticulin (residues 24-43) Synthetic peptide PEP-018 detects Ms Calreticulin (residues 24-43) in Not Applicable samples, and is validated for Control


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-018 is a 20 amino acid (amino acids 24-43) synthetic peptide whose sequence is found in the amino-terminal of mouse calreticulin. The sequence of this peptide is (amino to carboxy terminus): K-E-Q-F-L-D-G-D-A-W-T-N-R-W-V-E-S-K-H-K-C (The C-terminal cysteine has been added to facilitate conjugation and is not a part of the native mouse calreticulin sequence) This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and calreticulin, catalog # PA1-902A. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody. Reconstitute with 0.1 ml of distilled water.