Please login to your online account to display your discounted pricing

anti-Mouse Calreticulin (residues 24-43) Synthetic, Thermo Scientific™

Ms Calreticulin (residues 24-43) Synthetic peptide PEP-018 detects Ms Calreticulin (residues 24-43) in Not Applicable samples, and is validated for Control

$158.05 - $158.05

Specifications

Concentration 0.5mg/mL
Quantity 50μg
Format Purified
For Use With (Application) Control
Lyophilized Yes
View More Specs
Products
Catalog Number Mfr. No. Product Type Sequence Price Quantity    

PEP018

 
pierce biotechnology inc
PEP018
Mouse Calreticulin (Residues 24-43) Synthetic Peptide K-E -Q-F-L-D-G-D-A-W-T-N-R-W-V-E-S-K-H-K-C Each for $158.05

PEP017

 
pierce biotechnology inc
PEP017
Mouse Calreticulin (Residues 399-414) Synthetic Peptide C-D-E-K-E-E-D-E-E-E-S-P-G-Q-A-K-D Each for $158.05
Description & Specifications

Specifications

Concentration 0.5mg/mL
Quantity 50μg
Format Purified
For Use With (Application) Control
Lyophilized Yes
Purity >95%
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Ms Calreticulin (residues 24-43) Synthetic peptide PEP-018 detects Ms Calreticulin (residues 24-43) in Not Applicable samples, and is validated for Control

PEP-018 is a 20 amino acid (amino acids 24-43) synthetic peptide whose sequence is found in the amino-terminal of mouse calreticulin. The sequence of this peptide is (amino to carboxy terminus): K - E -Q - F - L - D - G - D - A - W - T - N - R - W - V - E - S - K - H - K - C (The C-terminal cysteine has been added to facilitate conjugation and is not a part of the native mouse calreticulin sequence)
Host Species: N/A
Cross Reactivity: Not Applicable
Formulation: 50μg purified Peptide (0.5mg/mL) no preservative;

PEP-018 detects Mouse Calreticulin (residues 24-43) in Not Applicable samples. PEP-018 has been successfully used in Control procedures. PEP-018