missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hamster Prion Protein, (residues 90-102) Synthetic Peptide, Invitrogen™

Hamster Prion Protein (residues 90-102) Synthetic Peptide, PEP-034, from Invitrogen™

Manufacturer:  Pierce Biotechnology Inc PEP034

Catalog No. PEP034


 

Specifications

Description

Hamster Prion Protein (residues 90-102) Synthetic Peptide, PEP-034, from Invitrogen.


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-034 is a 13 amino acid synthetic peptide whose sequence corresponds to amino acid residues 90-102 of hamster prion protein. The sequence of this peptide is (amino to carboxy terminus): G (90)-Q-G-G-G-T-H-N-Q-W-N-K-P (102)-G-G-C (Glycine and cysteine residues were added to facilitate coupling KLH). This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human prion protein, catalog # PA1-750. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody. Reconstitute with 0.1 ml of distilled water.
Description & Specifications

Specifications

>95%
0.5mg/mL
Purified
Yes
store at -20°C, Avoid Freeze/Thaw Cycles
50μg
Control
50μg purified peptide (0.5mg/mL) and no preservative
G(90)-Q-G-G-G-T-H-N-Q-W-N-K-P(102)-G-G-C
Hamster Prion Protein (Residues 90-102) Synthetic Peptide