Please login to your online account to display your discounted pricing

anti-Bovine Prion Protein (residues 146-159) Synthetic, Thermo Scientific™

Bv Prion Protein (residues 146-159) Synthetic peptide PEP-051 detects Bv Prion Protein (residues 146-159) in Not Applicable samples, and is validated for Control

$158.05 - $158.05

Specifications

Concentration 0.5mg/mL
Quantity 50μg
Format Purified
For Use With (Application) Control
Lyophilized Yes
View More Specs
Products
Catalog Number Mfr. No. Product Type Sequence Price Quantity    

PEP034

 
pierce biotechnology inc
PEP034
Hamster Prion Protein (Residues 90-102) Synthetic Peptide G(90)-Q-G-G-G-T-H-N-Q-W-N-K-P(102)-G-G-C Each for $158.05

PEP051

 
pierce biotechnology inc
PEP051
Bovine Prion Protein (Residues 146-159) Synthetic Peptide S146-R-P-L-I-H-F-G-S-D-Y-E-D-R159 Each for $158.05
Description & Specifications

Specifications

Concentration 0.5mg/mL
Quantity 50μg
Format Purified
For Use With (Application) Control
Lyophilized Yes
Purity >95%
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Bv Prion Protein (residues 146-159) Synthetic peptide PEP-051 detects Bv Prion Protein (residues 146-159) in Not Applicable samples, and is validated for Control

PEP-051 is a 14 amino acid synthetic peptide whose sequence corresponds to residues 146-159 of bovine Prion Protein. The sequence of this peptide is (amino to carboxy terminus): S146 - R - P - L - I - H - F - G - S - D - Y - E - D - R159
Host Species: N/A
Cross Reactivity: Not Applicable
Formulation: 50μg purified Peptide (0.5mg/mL) no preservative;

PEP-051 detects Bovine Prion Protein (residues 146-159) in Not Applicable samples. PEP-051 has been successfully used in Control procedures. PEP-051