Please login to your online account to display your discounted pricing

anti-Bovine Prion Protein (residues 146-159) Synthetic, Thermo Scientific™


Bv Prion Protein (residues 146-159) Synthetic peptide PEP-051 detects Bv Prion Protein (residues 146-159) in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  PEP051

 View more versions of this product

Catalog No. PEP051

  • / Each
 

Description & Specifications

Specifications

Formulation 50μg purified peptide (0.5mg/mL) and no preservative
For Use With (Application) Control
Format Purified
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Bovine Prion Protein (Residues 146-159) Synthetic Peptide
Purity >95%
Concentration 0.5mg/mL
Lyophilized Yes
Sequence S146-R-P-L-I-H-F-G-S-D-Y-E-D-R159
Quantity 50μg

Bv Prion Protein (residues 146-159) Synthetic peptide PEP-051 detects Bv Prion Protein (residues 146-159) in Not Applicable samples, and is validated for Control


Formulation: 50μg purified peptide (0.5mg/mL) and no preservative
PEP-051 is a 14 amino acid synthetic peptide whose sequence corresponds to residues 146-159 of bovine Prion Protein. The sequence of this peptide is (amino to carboxy terminus): S146-R-P-L-I-H-F-G-S-D-Y-E-D-R159 This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and prion protein, catalog # MA1-750. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∽70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 ml of distilled water.