Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Neuron Specific Enolase (human) Recombinant, Thermo Scientific™

Neuron Specific Enolase (Hu) Recombinant protein RP-75670 detects Neuron Specific Enolase (Hu) in Not Applicable samples, and is validated for Control and WB

$272.60 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Neuron Specific Enolase Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75668

 
pierce biotechnology inc
RP75668
10μg purified protein in PBS with 50% glycerol and no preservative 10μg Each for $272.60

PIRP75670

 
pierce biotechnology inc
RP75670
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Neuron Specific Enolase Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Neuron Specific Enolase (Hu) Recombinant protein RP-75670 detects Neuron Specific Enolase (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: Neuronal Specific Enolase, NSE

RP-75670 detects Neuron Specific Enolase (human) in Not Applicable samples. RP-75670 has been successfully used in Control and Western Blot procedures. RP-75670 The immunogen is N/A.