Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Myelin Fatty Acid Binding Protein (human) Recombinant, Thermo Scientific™

Myelin Fatty Acid Binding Protein (Hu) Recombinant protein RP-75557 detects Myelin Fatty Acid Binding Protein (Hu) in Not Applicable samples, and is validated for Control and WB

$288.55 - $448.05

Specifications

Cross Reactivity Human
Formulation 10μg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human Myelin Fatty Acid Binding Protein Recombinant Protein
Recombinant Yes
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PIRP75557

 
pierce biotechnology inc
RP75557
10μg Each for $288.55

PIRP75558

 
pierce biotechnology inc
RP75558
100μg Each for $448.05
Description & Specifications

Specifications

Cross Reactivity Human
Formulation 10μg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Name Human Myelin Fatty Acid Binding Protein Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Myelin Fatty Acid Binding Protein (Hu) Recombinant protein RP-75557 detects Myelin Fatty Acid Binding Protein (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 10μg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: Fatty Acid Binding Protein

RP-75557 detects Myelin Fatty Acid Binding Protein (human) in Not Applicable samples. RP-75557 has been successfully used in Control and Western Blot procedures. RP-75557 The immunogen is N/A.