Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Muscle-Cardiac FABP (human) Recombinant, Thermo Scientific™

Muscle-Cardiac FABP (Hu) Recombinant protein RP-75547 from Thermo Scientific™ specifically detects Muscle-Cardiac FABP (Hu) in Hu samples, and it is validated for Control and WB

$448.05 - $3,625.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Muscle Cardiac FABP Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75546

 
pierce biotechnology inc
RP75546
100μg purified protein (0.33mg/mL) in tris HCl with 50% glycerol and no preservative; pH 8.0 100μg Each for $448.05

PIRP75547

 
pierce biotechnology inc
RP75547
1mg purified protein (0.57mg/mL) in PBS with 50% glycerol and no preservative 1mg Each for $3,625.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Muscle Cardiac FABP Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Muscle-Cardiac FABP (Hu) Recombinant protein RP-75547 from Thermo Scientific™ specifically detects Muscle-Cardiac FABP (Hu) in Hu samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Human
Immunogen: N/A
Formulation: 1 mg purified Protein (0.57mg/mL) in PBS, with 50% glycerol and no preservative;
Synonyms: Fatty Acid Binding Protein
Uniprot Accession: P05413
Gene Symbol: FABP3
Entrez GeneID: 2170
RP-75547 detects Muscle-Cardiac FABP (human) in Human samples. RP-75547 has been successfully used in Control and Western Blot procedures. RP-75547 The immunogen is N/A.