Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Monoamine Oxidase B (human) Recombinant, Thermo Scientific™

Monoamine Oxidase B (Hu) Recombinant protein RP-75661 from Thermo Scientific™ detects Monoamine Oxidase B (Hu) in Not Applicable samples, and is validated for Control and WB

$785.90 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Monoamine Oxidase B Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75660

 
pierce biotechnology inc
RP75660
100μg purified protein (0.11mg/mL) in TBS with 50% glycerol, 1mM EDTA and no preservative; pH 8 100μg Each for $785.90

PIRP75661

 
pierce biotechnology inc
RP75661
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Monoamine Oxidase B Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Monoamine Oxidase B (Hu) Recombinant protein RP-75661 from Thermo Scientific™ detects Monoamine Oxidase B (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: MAO b

RP-75661 detects Monoamine Oxidase B (human) in Not Applicable samples. RP-75661 has been successfully used in Control and Western Blot procedures. RP-75661 The immunogen is N/A.