Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Mitochondrial Glycerol-3-PO4 Dehydrogenase Recombinant, Thermo Scientific™

Mitochondrial Glycerol-3-PO4 Dehydrogenase Recombinant protein RP-75658 detects Mitochondrial Glycerol-3-PO4 Dehydrogenase in Not Applicable samples, and is validated for Control and WB

$288.55 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Mitochondrial Glycerol-3-PO4 Dehydrogenase Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75656

 
pierce biotechnology inc
RP75656
10μg purified protein in PBS with 50% glycerol and no preservative 10μg Each for $288.55

PIRP75657

 
pierce biotechnology inc
RP75657
100μg purified protein in PBS with 50% glycerol and no preservative 100μg Each for $797.50

PIRP75658

 
pierce biotechnology inc
RP75658
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Mitochondrial Glycerol-3-PO4 Dehydrogenase Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Mitochondrial Glycerol-3-PO4 Dehydrogenase Recombinant protein RP-75658 detects Mitochondrial Glycerol-3-PO4 Dehydrogenase in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: Mitochondrial Glycerol-3-Phosphate Dehydrogenase

RP-75658 detects Mitochondrial Glycerol-3-PO4 Dehydrogenase in Not Applicable samples. RP-75658 has been successfully used in Control and Western Blot procedures. RP-75658 The immunogen is N/A.