Please login to your online account to display your discounted pricing

anti-Luteinizing Hormone Releasing Hormone Synthetic, Thermo Scientific™

Luteinizing Hormone Releasing Hormone Synthetic peptide PEP-10933 detects Luteinizing Hormone Releasing Hormone in Not Applicable samples, and is validated for Control

$685.85 - $797.50

Specifications

Format Purified
Lyophilized Yes
Sequence Pyr-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. For Use With (Application) Storage Requirements Quantity Price Quantity    

PIPEP10933

 
pierce biotechnology inc
PEP10933
Control store at -20°C, Avoid Freeze/Thaw Cycles 10mg Each for $685.85

PIPEP10934

 
pierce biotechnology inc
PEP10934
Control Store at -80°C, Avoid Freeze/Thaw Cycles 25mg Each for $797.50
Description & Specifications

Specifications

Format Purified
Lyophilized Yes
Sequence Pyr-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2

Luteinizing Hormone Releasing Hormone Synthetic peptide PEP-10933 detects Luteinizing Hormone Releasing Hormone in Not Applicable samples, and is validated for Control


Host Species: N/A
Cross Reactivity: Not Applicable
Clone: N/A
Immunogen: N/A
Formulation: 10 mg purified Peptide no preservative;
Synonyms: LHRH, GnRH, Lutropin

PEP-10933 detects Luteinizing Hormone Releasing Hormone in Not Applicable samples. PEP-10933 has been successfully used in Control procedures. PEP-10933 The immunogen is N/A.