Please login to your online account to display your discounted pricing

anti-Luteinizing Hormone Releasing Hormone Synthetic, Thermo Scientific™


Luteinizing Hormone Releasing Hormone Synthetic peptide PEP-10933 detects Luteinizing Hormone Releasing Hormone in Not Applicable samples, and is validated for Control

Manufacturer: pierce biotechnology inc  PEP10934

 View more versions of this product

 Disclaimers

in vitro research only.

Catalog No. PIPEP10934

  • / Each
 

Description & Specifications

Specifications

Formulation 25mg purified peptide and no preservative
For Use With (Application) Control
Format Purified
Gene Alias LHRH, GnRH, Lutropin
Storage Requirements Store at -80°C, Avoid Freeze/Thaw Cycles
Product Type Luteinizing Hormone Releasing Hormone Synthetic Peptide
Sequence Pyr-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2
Lyophilized Yes
Quantity 25mg

Luteinizing Hormone Releasing Hormone Synthetic peptide PEP-10933 detects Luteinizing Hormone Releasing Hormone in Not Applicable samples, and is validated for Control


Formulation: 25mg purified peptide and no preservative
PEP-10934 is a synthetic peptide suitable for use as a control in ELISA and Western blot applications. PEP-10934 has the amino acid sequence: Pyr-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2.