Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-L-Selectin (human) Recombinant, Thermo Scientific™

L-Selectin (Hu) Recombinant protein RP-75649 from Thermo Scientific™ specifically detects L-Selectin (Hu) in Not Applicable samples, and it is validated for Control and WB

$288.55 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human L-Selectin Recombinant Protein
Recombinant Yes
Source Human
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75647

 
pierce biotechnology inc
RP75647
10μg purified protein in PBS with 50% glycerol and no preservative 10μg Each for $288.55

PIRP75649

 
pierce biotechnology inc
RP75649
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00

PIRP75648

 
pierce biotechnology inc
RP75648
100μg purified protein in PBS with 50% glycerol and no preservative 100μg Each for $870.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human L-Selectin Recombinant Protein
Recombinant Yes
Source Human
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

L-Selectin (Hu) Recombinant protein RP-75649 from Thermo Scientific™ specifically detects L-Selectin (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75649 detects L-Selectin (human) in Not Applicable samples. RP-75649 has been successfully used in Control and Western Blot procedures. RP-75649 The immunogen is N/A.