Please login to your online account to display your discounted pricing

anti-Intestinal Fatty Acid Binding Protein (human) Recombinant, Thermo Scientific™


Intestinal Fatty Acid Binding Protein (Hu) Recombinant protein RP-75551 detects Intestinal Fatty Acid Binding Protein (Hu) in Not Applicable samples, and is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75552

 View more versions of this product

 Disclaimers

in vitro research only.

Catalog No. PIRP75552

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg purified protein in PBS with 50% glycerol and no preservative
For Use With (Application) Control, Western Blot
Conjugate Unlabeled
Host Species E. Coli
Format Purified
Gene Alias Fatty Acid Binding Protein
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Name Human Intestinal Fatty Acid Binding Protein Recombinant Protein
Quantity 100μg
Recombinant Yes
Source E. coli
Cross Reactivity Human

Intestinal Fatty Acid Binding Protein (Hu) Recombinant protein RP-75551 detects Intestinal Fatty Acid Binding Protein (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Formulation: 100μg purified protein in PBS with 50% glycerol and no preservative
RP-75552 contains purified recombinant human intestinal FABP protein with an N-terminal hexahistidine tag. RP-75552 may be used as a positive control in many immunological assays.