Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Interferon Inducible Protein (human) Recombinant, Thermo Scientific™

Interferon Inducible Protein (Hu) Recombinant protein RP-75646 detects Interferon Inducible Protein (Hu) in Not Applicable samples, and is validated for Control and WB

$288.55 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Interferon Inducible Protein Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75645

 
pierce biotechnology inc
RP75645
100μg purified protein in PBS with 50% glycerol and no preservative 100μg Each for $797.50

PIRP75644

 
pierce biotechnology inc
RP75644
10μg purified protein in 10mM tris HCl with 50% glycerol, 1mM EDTA and no preservative; pH 8.0 10μg Each for $288.55

PIRP75646

 
pierce biotechnology inc
RP75646
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Interferon Inducible Protein Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Interferon Inducible Protein (Hu) Recombinant protein RP-75646 detects Interferon Inducible Protein (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75646 detects Interferon Inducible Protein (human) in Not Applicable samples. RP-75646 has been successfully used in Control and Western Blot procedures. RP-75646 The immunogen is N/A.