Please login to your online account to display your discounted pricing

anti-Interferon Inducible Protein (human) Recombinant, Thermo Scientific™


Interferon Inducible Protein (Hu) Recombinant protein RP-75646 detects Interferon Inducible Protein (Hu) in Not Applicable samples, and is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75646

 View more versions of this product

 Disclaimers

in vitro research only.

Catalog No. PIRP75646

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control, Western Blot
Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
Conjugate Unlabeled
Host Species E. Coli
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Name Human Interferon Inducible Protein Recombinant Protein
Recombinant Yes
Source E. coli
Quantity 1mg
Cross Reactivity Human

Interferon Inducible Protein (Hu) Recombinant protein RP-75646 detects Interferon Inducible Protein (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Formulation: 1mg purified protein in PBS with 50% glycerol and no preservative
RP-75646 contains purified recombinant human IFN Inducible Protein protein with an N-terminal hexahistidine tag. RP-75646 may be used as a positive control in many immunological assays.