Please login to your online account to display your discounted pricing

anti-Interferon gamma (human) Recombinant, Thermo Scientific™


Interferon gamma (Hu) Recombinant protein RP-75631 from Thermo Scientific™ detects Interferon gamma (Hu) in Not Applicable samples, and is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75629

 View more versions of this product

 Disclaimers

in vitro research only.

Catalog No. PIRP75629

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control, Western Blot
Formulation 10μg purified protein (0.23mg/mL) in 20mM tris HCl with 50% glycerol and no preservative; pH 8.0
Host Species E. Coli
Conjugate Unlabeled
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Immunogen N/A
Name Human Interferon gamma Recombinant Protein
Source E. coli
Quantity 10μg
Cross Reactivity Human
Recombinant Yes
Concentration 0.1mg/mL

Interferon gamma (Hu) Recombinant protein RP-75631 from Thermo Scientific™ detects Interferon gamma (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Formulation: 10μg purified protein (0.23mg/mL) in 20mM tris HCl with 50% glycerol and no preservative; pH 8.0
RP-75629 contains purified recombinant human IFN gamma protein with an N-terminal hexahistidine tag. RP-75629 may be used as a positive control in many immunological assays.