Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Interferon gamma (human) Recombinant, Thermo Scientific™

Interferon gamma (Hu) Recombinant protein RP-75631 from Thermo Scientific™ detects Interferon gamma (Hu) in Not Applicable samples, and is validated for Control and WB

$272.60 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Interferon gamma Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75630

 
pierce biotechnology inc
RP75630
100μg purified protein in 20mM tris HCl with 50% glycerol and no preservative; pH 8 100μg Each for $759.80

PIRP75631

 
pierce biotechnology inc
RP75631
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00

PIRP75629

 
pierce biotechnology inc
RP75629
10μg purified protein (0.23mg/mL) in 20mM tris HCl with 50% glycerol and no preservative; pH 8.0 10μg Each for $272.60
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Interferon gamma Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Interferon gamma (Hu) Recombinant protein RP-75631 from Thermo Scientific™ detects Interferon gamma (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75631 detects Interferon gamma (human) in Not Applicable samples. RP-75631 has been successfully used in Control and Western Blot procedures. RP-75631 The immunogen is N/A.