Please login to your online account to display your discounted pricing

anti-Interferon gamma (human) Recombinant, Thermo Scientific™


Interferon gamma (Hu) Recombinant protein RP-75631 from Thermo Scientific™ detects Interferon gamma (Hu) in Not Applicable samples, and is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75630

 View more versions of this product

 Disclaimers

in vitro research only.

Catalog No. PIRP75630

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) Control, Western Blot
Formulation 100μg purified protein in 20mM tris HCl with 50% glycerol and no preservative; pH 8
Host Species E. Coli
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Name Human Interferon gamma Recombinant Protein
Recombinant Yes
Source E. coli
Concentration 0.1mg/mL
Cross Reactivity Human
Quantity 100μg

Interferon gamma (Hu) Recombinant protein RP-75631 from Thermo Scientific™ detects Interferon gamma (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Formulation: 100μg purified protein in 20mM tris HCl with 50% glycerol and no preservative; pH 8
RP-75630 contains purified recombinant human IFN gamma protein with an N-terminal hexahistidine tag. RP-75630 may be used as a positive control in many immunological assays.