Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Integrin-Linked Kinase (human) Recombinant, Thermo Scientific™

Integrin-Linked Kinase (Hu) Recombinant protein RP-75640 detects Integrin-Linked Kinase (Hu) in Not Applicable samples, and is validated for Control and WB

$326.25 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Integrin-Linked Kinase Recombinant Protein
Purity >95%
Recombinant Yes
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Molecular Weight Quantity Price Quantity    

PIRP75638

 
pierce biotechnology inc
RP75638
10μg purified protein (0.05mg/mL) in Laemmli and no preservative; pH 6.8 55.92 10μg Each for $326.25

PIRP75639

 
pierce biotechnology inc
RP75639
100μg purified protein in PBS with 50% glycerol and no preservative ∽59 100μg Each for $797.50

PIRP75640

 
pierce biotechnology inc
RP75640
1mg purified protein in PBS with 50% glycerol and no preservative ∽59 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human Integrin-Linked Kinase Recombinant Protein
Purity >95%
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Integrin-Linked Kinase (Hu) Recombinant protein RP-75640 detects Integrin-Linked Kinase (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75640 detects Integrin-Linked Kinase (human) in Not Applicable samples. RP-75640 has been successfully used in Control and Western Blot procedures. RP-75640 The immunogen is N/A.