Please login to your online account to display your discounted pricing

anti-Integrin-Linked Kinase (human) Recombinant, Thermo Scientific™


Integrin-Linked Kinase (Hu) Recombinant protein RP-75640 detects Integrin-Linked Kinase (Hu) in Not Applicable samples, and is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75639

 View more versions of this product

 Disclaimers

in vitro research only.

Catalog No. PIRP75639

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) Control, Western Blot
Formulation 100μg purified protein in PBS with 50% glycerol and no preservative
Host Species E. Coli
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Purification Method SDS-PAGE
Molecular Weight ∽59
Purity >95%
Quantity 100μg
Recombinant Yes
Source E. coli
Name Human Integrin-Linked Kinase Recombinant Protein
Cross Reactivity Human

Integrin-Linked Kinase (Hu) Recombinant protein RP-75640 detects Integrin-Linked Kinase (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Formulation: 100μg purified protein in PBS with 50% glycerol and no preservative
RP-75639 contains a purified 452 amino acid fragment (aa1-452) of mature human ILK1. RP-75639 may be used as a positive control in many immunological assays. This protein has a molecular weight of ∽59 kDa. Purity >95% by SDS-PAGE.