Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Inhibin a (human) Recombinant, Thermo Scientific™

Inhibin a (Hu) Recombinant protein RP-75634 from Thermo Scientific™ specifically detects Inhibin a (Hu) in Not Applicable samples, and it is validated for Control and WB

$303.05 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Molecular Weight 19.2
Name Human Inhibit a Recombinant Protein
Recombinant Yes
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75632

 
pierce biotechnology inc
RP75632
10μg purified protein (0.05mg/mL) in Laemmli and no preservative; pH 6.8 10μg Each for $303.05

PIRP75633

 
pierce biotechnology inc
RP75633
100μg purified protein in tris HCl with 50% glycerol, 5mM EDTA and no preservative; pH 8 100μg Each for $785.90

PIRP75634

 
pierce biotechnology inc
RP75634
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Molecular Weight 19.2
Name Human Inhibit a Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Inhibin a (Hu) Recombinant protein RP-75634 from Thermo Scientific™ specifically detects Inhibin a (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75634 detects Inhibin a (human) in Not Applicable samples. RP-75634 has been successfully used in Control and Western Blot procedures. RP-75634 The immunogen is N/A.