Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Inhibin beta A (human) Recombinant, Thermo Scientific™

Inhibin beta A (Hu) Recombinant protein RP-75637 from Thermo Scientific™ specifically detects Inhibin beta A (Hu) in Not Applicable samples, and is validated for Control and WB

$326.25 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Molecular Weight 17.47
Name Human Inhibit beta A Chain Recombinant Protein
Recombinant Yes
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75636

 
pierce biotechnology inc
RP75636
100μg purified protein (0.13mg/mL) in 0.02M tris with 50% glycerol, 1mM EDTA and no preservative 100μg Each for $800.40

PIRP75637

 
pierce biotechnology inc
RP75637
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00

PIRP75635

 
pierce biotechnology inc
RP75635
10μg purified protein (0.05mg/mL) in Laemmli and no preservative; pH 6.8 10μg Each for $326.25
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Molecular Weight 17.47
Name Human Inhibit beta A Chain Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Inhibin beta A (Hu) Recombinant protein RP-75637 from Thermo Scientific™ specifically detects Inhibin beta A (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75637 detects Inhibin beta A (human) in Not Applicable samples. RP-75637 has been successfully used in Control and Western Blot procedures. RP-75637 The immunogen is N/A.