Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-Ileal Fatty Acid Binding Protein (human) Recombinant, Thermo Scientific™

Ileal Fatty Acid Binding Protein (Hu) Recombinant protein RP-75560 detects Ileal Fatty Acid Binding Protein (Hu) in Not Applicable samples, and is validated for Control and WB

$288.55 - $543.75

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Molecular Weight ∽18.87
Name Human Ileal Fatty Acid Binding Protein Recombinant Protein
Recombinant Yes
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75561

 
pierce biotechnology inc
RP75561
100μg purified protein (0.5mg/mL) in tris HCl with 50% glycerol and no preservative 100μg Each for $543.75

PIRP75560

 
pierce biotechnology inc
RP75560
10μg purified protein in PBS with 50% glycerol and no preservative 10μg Each for $288.55
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Molecular Weight ∽18.87
Name Human Ileal Fatty Acid Binding Protein Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

Ileal Fatty Acid Binding Protein (Hu) Recombinant protein RP-75560 detects Ileal Fatty Acid Binding Protein (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 10μg purified Protein in PBS, with 50% glycerol and no preservative;
Synonyms: Fatty Acid Binding Protein

RP-75560 detects Ileal Fatty Acid Binding Protein (human) in Not Applicable samples. RP-75560 has been successfully used in Control and Western Blot procedures. RP-75560 The immunogen is N/A.