Please login to your online account to display your discounted pricing

anti-IL-8 (human) Recombinant, Thermo Scientific™


IL-8 (human) Recombinant protein RP-75610 from Thermo Scientific™ specifically detects IL-8 (human) in Not Applicable samples, and it is validated for Control and Western Blot

Manufacturer: pierce biotechnology inc  RP75609

 View more versions of this product

 Disclaimers

in vitro research only.

Catalog No. PIRP75609

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Control, Western Blot
Formulation 100μg purified protein in TBS with 50% glycerol and no preservative; pH 8
Conjugate Unlabeled
Host Species E. Coli
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Name Human IL-8 Recombinant Protein
pH Range 8
Quantity 100μg
Recombinant Yes
Source E. coli
Cross Reactivity Human

IL-8 (human) Recombinant protein RP-75610 from Thermo Scientific™ specifically detects IL-8 (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Formulation: 100μg purified protein in TBS with 50% glycerol and no preservative; pH 8
RP-75609 contains a purified 77 amino acid fragment (aa23-99) corresponding to the mature human IL-8 protein with an N-terminal hexahistidine tag. RP-75609 may be used as a positive control in many immunological assays.