Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-IL-8 (human) Recombinant, Thermo Scientific™

IL-8 (human) Recombinant protein RP-75610 from Thermo Scientific™ specifically detects IL-8 (human) in Not Applicable samples, and it is validated for Control and Western Blot

$269.70 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human IL-8 Recombinant Protein
pH Range 8
Recombinant Yes
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75609

 
pierce biotechnology inc
RP75609
100μg purified protein in TBS with 50% glycerol and no preservative; pH 8 100μg Each for $759.80

PIRP75610

 
pierce biotechnology inc
RP75610
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00

PIRP75608

 
pierce biotechnology inc
RP75608
10μg purified protein in TBS with 50% glycerol and no preservative; pH 8 10μg Each for $269.70
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human IL-8 Recombinant Protein
pH Range 8
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

IL-8 (human) Recombinant protein RP-75610 from Thermo Scientific™ specifically detects IL-8 (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75610 detects IL-8 (human) in Not Applicable samples. RP-75610 has been successfully used in Control and Western Blot procedures. RP-75610 The immunogen is N/A.