missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Human IL-6R (soluble) Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP75602
Description
IL-6 Soluble Receptor (human) Recombinant Protein, RP-75602, from Invitrogen.
The IL-6 receptor complex is composed of two membrane glycoproteins: the low affinity receptor and the signaltransducing component. The soluble form of IL6R is found in the urine of healthy adult humans and the serum of HIV positive individuals, as well as in the cell culture supernatants of stimulated PBMCâs. This soluble form of IL6R results from either proteolytic cleavage from the membrane, or an isoform derived splice variant.The IL-6 receptor complex is composed of two membrane glycoproteins: the low affinity receptor and the signaltransducing component. The soluble form of IL6R is found in the urine of healthy adult humans and the serum of HIV positive individuals, as well as in the cell culture supernatants of stimulated PBMCâs. This soluble form of IL6R results from either proteolytic cleavage from the membrane, or an isoform derived splice variant. RP-75602 contains purified recombinant human IL-6 Soluble Receptor protein with an N-terminal hexahistidine tag. RP-75602 may be used as a positive control in many immunological assays.Specifications
P08887 | |
Control, Western Blot | |
3570 | |
6.8 | |
E. coli | |
RUO | |
IL6R | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
0.05mg/mL | |
Laemmli with no preservative; pH 6.8 | |
Human IL-6 Soluble Receptor | |
10μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
Human IL-6 Soluble Receptor | |
Human | |
Hu | |
Recombinant | |
Recombinant |