Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-IL-6 Soluble Receptor (human) Recombinant, Thermo Scientific™

IL-6 Soluble Receptor (Hu) Recombinant protein RP-75604 from Thermo Scientific™ detects IL-6 Soluble Receptor (Hu) in Not Applicable samples, and is validated for Control and WB

$303.05 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human IL-6 Soluble Receptor Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro research only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75604

 
pierce biotechnology inc
RP75604
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00

PIRP75602

 
pierce biotechnology inc
RP75602
10μg purified protein (0.05mg/mL) in Laemmli and no preservative; pH 6.8 10μg Each for $303.05
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human IL-6 Soluble Receptor Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

IL-6 Soluble Receptor (Hu) Recombinant protein RP-75604 from Thermo Scientific™ detects IL-6 Soluble Receptor (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75604 detects IL-6 Soluble Receptor (human) in Not Applicable samples. RP-75604 has been successfully used in Control and Western Blot procedures. RP-75604 The immunogen is N/A.