Please login to your online account to display your discounted pricing

anti-IL-3 (human) Recombinant, Thermo Scientific™


IL-3 (human) Recombinant protein RP-75595 from Thermo Scientific™ specifically detects IL-3 (human) in Not Applicable samples, and it is validated for Control and Western Blot

Manufacturer: pierce biotechnology inc  RP75594

 View more versions of this product

 Disclaimers

in vitro research only.

Catalog No. PIRP75594

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) Control, Western Blot
Formulation 100μg purified protein in TBS with 50% glycerol and no preservative
Host Species E. Coli
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Name Human IL-3 Recombinant Protein
Quantity 100μg
Recombinant Yes
Source E. coli
Cross Reactivity Human

IL-3 (human) Recombinant protein RP-75595 from Thermo Scientific™ specifically detects IL-3 (human) in Not Applicable samples, and it is validated for Control and Western Blot


Host Species: E. Coli
Formulation: 100μg purified protein in TBS with 50% glycerol and no preservative
RP-75594 contains purified recombinant human IL-3 protein with an N-terminal hexahistidine tag. RP-75594 may be used as a positive control in many immunological assays.