Please login to your online account to display your discounted pricing

Thermo Scientific™ anti-IL-2 Receptor alpha (human) Recombinant, Thermo Scientific™

IL-2 Receptor alpha (Hu) Recombinant protein RP-75592 from Thermo Scientific™ detects IL-2 Receptor alpha (Hu) in Not Applicable samples, and is validated for Control and WB

$288.55 - $3,770.00

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human IL-2 Receptor alpha Recombinant Protein
Recombinant Yes
Source E. coli
View More Specs

 Disclaimers

in vitro reserach only.

Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

PIRP75590

 
pierce biotechnology inc
RP75590
10μg purified protein in PBS with 50% glycerol and no preservative 10μg Each for $288.55

PIRP75591

 
pierce biotechnology inc
RP75591
100μg purified protein in PBS with 50% glycerol and no preservative; pH 7.4 100μg Each for $870.00

PIRP75592

 
pierce biotechnology inc
RP75592
1mg purified protein in PBS with 50% glycerol and no preservative 1mg Each for $3,770.00
Description & Specifications

Specifications

Cross Reactivity Human
For Use With (Application) Control, Western Blot
Name Human IL-2 Receptor alpha Recombinant Protein
Recombinant Yes
Source E. coli
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles

IL-2 Receptor alpha (Hu) Recombinant protein RP-75592 from Thermo Scientific™ detects IL-2 Receptor alpha (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Cross Reactivity: Not Applicable
Immunogen: N/A
Formulation: 1 mg purified Protein in PBS, with 50% glycerol and no preservative;

RP-75592 detects IL-2 Receptor alpha (human) in Not Applicable samples. RP-75592 has been successfully used in Control and Western Blot procedures. RP-75592 The immunogen is N/A.