Please login to your online account to display your discounted pricing

anti-IL-2 Receptor alpha (human) Recombinant, Thermo Scientific™


IL-2 Receptor alpha (Hu) Recombinant protein RP-75592 from Thermo Scientific™ detects IL-2 Receptor alpha (Hu) in Not Applicable samples, and is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75592

 View more versions of this product

 Disclaimers

in vitro reserach only.

Catalog No. PIRP75592

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) Control, Western Blot
Formulation 1mg purified protein in PBS with 50% glycerol and no preservative
Host Species E. Coli
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Name Human IL-2 Receptor alpha Recombinant Protein
Quantity 1mg
Recombinant Yes
Source E. coli
Cross Reactivity Human

IL-2 Receptor alpha (Hu) Recombinant protein RP-75592 from Thermo Scientific™ detects IL-2 Receptor alpha (Hu) in Not Applicable samples, and is validated for Control and WB


Host Species: E. Coli
Formulation: 1mg purified protein in PBS with 50% glycerol and no preservative
RP-75592 contains purified recombinant human IL-2 Receptor alpha protein with an N-terminal hexahistidine tag. RP-75592 may be used as a positive control in many immunological assays.