Please login to your online account to display your discounted pricing

anti-IL-12 p40 (human) Recombinant, Thermo Scientific™


IL-12 p40 (Hu) Recombinant protein RP-75619 from Thermo Scientific™ specifically detects IL-12 p40 (Hu) in Not Applicable samples, and it is validated for Control and WB

Manufacturer: pierce biotechnology inc  RP75618

 View more versions of this product

 Disclaimers

in vitro research only

Catalog No. PIRP75618

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
For Use With (Application) Control, Western Blot
Formulation 100μg purified protein in PBS with 50% glycerol and no preservative
Host Species E. Coli
Immunogen N/A
Storage Requirements store at -20°C, Avoid Freeze/Thaw Cycles
Format Purified
Recombinant Yes
Source E. coli
Molecular Weight 16.75
Name Human IL-12 p40 Recombinant Protein
Quantity 100μg
Cross Reactivity Human

IL-12 p40 (Hu) Recombinant protein RP-75619 from Thermo Scientific™ specifically detects IL-12 p40 (Hu) in Not Applicable samples, and it is validated for Control and WB


Host Species: E. Coli
Formulation: 100μg purified protein in PBS with 50% glycerol and no preservative
RP-75618 contains purified recombinant human IL-12 p40 protein with an N-terminal hexahistidine tag. RP-75618 may be used as a positive control in many immunological assays.